Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium chloride, ACS, 99% min, Thermo Scientific Chemicals

Catalog Number aa3621736
Click to view available options
Quantity:
500 g
2 kg
7447-41-8
ClLi
42.39
MFCD00011078
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
433294
CHEBI:48607
lithium(1+) chloride
[Li+].[Cl-]
7447-41-8
ClLi
42.39
MFCD00011078
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
433294
CHEBI:48607
lithium(1+) chloride
[Li+].[Cl-]

In air-conditioning, welding and soldering flux, dry batteries, heat-exchange media, salt baths, and desiccants.Lithium chloride is useful for the production of lithium metal, and for the generation of Mn(0) species which can be used in free radical cyclizations. It can serve as a flame colorant to generate dark red flames, a brazing flux for aluminum in automobiles, a hygrometer, and a desiccant for drying air streams. On exposure to air, it becomes a solution with the concentration directly related to relative humidity of the atmosphere, and hence serves as a relative humidity standard in calibrating hygrometers. Apart from being a source of chloride, it serves as an additive in the Stille reaction in organic synthesis, and to precipitate RNA from cellular extracts. Being biologically significant, it finds applications in a wide variety of assays to study cell-fate and neurobiology. Lithium chloride has been found to inhibit virus infection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1325°C to 1360°C
Melting Point 605°C
Physical Form Granules
Quantity 500 g
Assay Percent Range 99%
Solubility Information Very soluble in water,alcohol,ether,pyridine,and nitrobenzene.
Formula Weight 42.39
Grade ACS Reagent
Sensitivity Hygroscopic
Density 2.068 g/mL
Chemical Name or Material Lithium chloride
Show More Show Less
Hazard Category H302-H315-H319
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
EINECSNumber 231-212-3
RTECSNumber OJ5950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit