Learn More
Thermo Scientific™ LN2 Low-Level Alarms
Add an extra level of protection to the lab in the event of excessive liquid nitrogen loss during dewar storage with LN2 Low-Level Alarms.
Supplier: Thermo Scientific™ 8130
Description
Add an extra level of protection to the laboratory in the event of excessive liquid nitrogen loss during dewar storage with Thermo Scientific™ LN2 Low-Level Alarms, for use with any standard-sized LN2 container.
Low level alarm (120V) with sensor for any standard-sized LN2 container.Includes:
Dry contacts for remote alarm
Specifications
Low-level alarm with dry contacts for connection to telephone dialing system or remote alarm, customer installed | |
Any Standard-Sized LN2 Container | |
120 V |
LN2 Low-Level Alarm | |
Alarm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.