Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Matrix™ Filtered 384 Format Pipette Tips

Eliminate pipetter barrel contamination with Thermo Scientific™ Matrix™ Filtered Pipette Tips.

Specifications

Filtered Filtered
Sterility Sterile
Format Rack
For Use With (Equipment) Equalizer 384, Thermo Scientific Matrix Impact 16 Channel
Product Line Matrix
View More Specs

Products 1
Catalog Number Mfr. No. Volume (Metric) For Use With (Equipment) Format Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) For Use With (Equipment) Format Price Quantity & Availability  
14-387-199
View Documents
Thermo Scientific™
7445
125 μL Equalizer 384, Thermo Scientific Matrix Impact 16 Channel Rack Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

Eliminate pipetter barrel contamination with Thermo Scientific™ Matrix™ Filtered Pipette Tips. Rigorously tested to ensure optimum performance, these filtered tips eliminate the risk of cross-contamination from aerosols created while pipetting genomic material, bacterial samples or other precious samples.

Benefits:
  • Stringent testing guarantees filter tips that effectively block aerosols.
  • Hydrophobic polyethylene filter material ensures no particulates contaminate samples.
  • Inadvertent contact with filter will not contaminate sample or result in sample loss.
  • Easy to open packaging maintains product sterility while providing easy access.
  • Hinged lid allows rack to be opened without laying lid on benchtop, helping maintain sterility.
  • Racks are sealed in thick polyethylene bag for sterility, which is easy to open with simple snap-and-tear packaging.

Stringent Filter Testing:

  • Filter must meet certain porosity requirements and must form a complete barrier across face of tip.
  • To ensure defects are not present, two specific tests are performed: mercury intrusion and particle count testing.
  • Mercury Intrusion Testing (QC 13.1 - QC 13.8) ensures the filter itself meets specific aerosol blocking requirements.
  • Particle Count Testing (Test Protocol QAS-0100) ensures filter is effective at blocking aerosols after installation into tip; also ensures proper shape and fit of filter.
  • Every lot of tips is tested by drawing a vacuum of aerosols against the filter and ensuring particles do not pass through.

Recommended for:

  • Bacteriology
  • Gel loading
  • PCR setup
  • Preventing cross-contamination during routine pipetting
Specifications

Specifications

Filtered
Rack
Matrix
Sterile
Equalizer 384, Thermo Scientific Matrix Impact 16 Channel
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit