Learn More
Thermo Scientific™ MBTFA Acylation Reagent
Trifluoroacylate primary and secondary amines, hydroxyl and thiol groups, and carbohydrates with the Thermo Scientific™ MBTFA Acylation Reagent.
$365.00
Specifications
Standard Components | N-Methyl-bis(trifluoroacetamide) |
---|---|
Storage | -20°C |
For Use With | GC |
Hazards | Corrosive, Flammable |
Standard Concentration | 98 to 100% |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PI49700
|
Thermo Scientific™
TS49700 |
10 x 1 mL ampules |
Pack of 10 for $365.00
|
|
|||||
TS49701
|
Thermo Scientific™
TS49701 |
5 g |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | |||||
PI49703
|
Thermo Scientific™
TS49703 |
25 mL |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | |||||
PI49704
|
Thermo Scientific™
TS49704 |
100 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Trifluoroacylate primary and secondary amines, hydroxyl and thiol groups, and carbohydrates with the Thermo Scientific™ MBTFA Acylation Reagent. The principal byproduct N-methyltrifluoroacetamine is stable, volatile, and does not present problems in subsequent GC. MBTFA produces very volatile derivatives of carbohydrates and can be used to selectively acylate amines in the presence of hydroxyl and carboxyl groups that have been protected by silylation.
- Reacts under nonacidic conditions
- Principle byproduct from the derivatization reaction is N-methyltrifluoroacetamine, which is stable, volatile and does not present problems in subsequent GC
- Produces very volatile derivatives of carbohydrates
- Can be used to selectively acylate amines in the presence of hydroxyl and carboxyl groups that have been protected by silylation
Specifications
N-Methyl-bis(trifluoroacetamide) | |
GC | |
98 to 100% |
-20°C | |
Corrosive, Flammable |
For Research Use Only. Not for use in diagnostic procedures.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.