Learn More
Mettler Toledo™ Evaporation Trap for XP/XS Balances
Converts balances into pipette calibration station.
Supplier: Mettler Toledo™ 11140043
Description
Mettler Toledo evaporation traps mount on their respective balances to provide a perfect setup for pipette testing within minutes.
Specifications
XP/XS Analytical Balance | |
Evaporation Trap |