Learn More
Mettler Toledo™ Tweezers for Analytical and Calibration Weights
Accessories for Weights. Ergonomic Weight Handling - Ergonomic tweezers and weight forks are made of clean room suitable materials and allow professional weight handling and balance testing.
$110.24 - $153.17
Specifications
Product Type Tweezers
Material Metal with Carbon Reinforced Nylon
Quantity Each
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) Price Quantity & Availability  
01-915-379
View Documents
Mettler Toledo™
15901
1g-1kg Weights
Each for $153.17
Add to Cart
01-915-378
View Documents
Mettler Toledo™
15900
1mg to 50g Weights
Each for $110.24
Add to Cart
Description
Description
Metal with carbon reinforced nylon.
Specifications
Specifications
Tweezers
Metal with Carbon Reinforced Nylon
Each