Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minigrip™ SPECI-ZIP™ Reclosable Red Biohazard Bags

With black STAT and biohazard warning

$128.00 - $181.00

Specifications

Product Type Reclosable Biohazard Bag
Color Red
For Use With (Application) Designed with info pocket, temperature indicators and tear line
Includes Black STAT/ biohazard warning
View More Specs

Products 3
Catalog Number Mfr. No. Width (English) Width (Metric) Height (English) Height (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Width (English) Width (Metric) Height (English) Height (Metric) Price Quantity & Availability  
22-310-410
View Documents
Minigrip™
IP810RSTAT
8 in. 20.3 cm 10 in. 25.4 cm
Case of 1000 for $181.00
Edge
Add to Cart
 
22-310-409
View Documents
Minigrip™
IP69RSTAT
6 in. 15.24 cm 9 in. 22.86 cm
Case of 1000 for $130.00
Add to Cart
 
22-310-096
View Documents
Minigrip™
IP69RSTAT2
6 in. 15.2 cm 9 in. 22.86 cm
Case of 1000 for $128.00
Add to Cart
 
Description

Description

  • Designed with info pocket (three wall) and tear line
  • Bilingual in English and Spanish

Order Info

Case of 1000 in packs of 100.

Specifications

Specifications

Reclosable Biohazard Bag
Red
Designed with info pocket, temperature indicators and tear line
Black STAT/ biohazard warning
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit