Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Mouse CCL3/MIP-1 alpha Quantikine ELISA Kit
Mouse CCL3/MIP-1 alpha Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
$922.20 - $4199.20
Specifications
Assay Range | 4.70 - 300pg/mL |
---|---|
Assay Sensitivity | 1.5pg/mL |
Conjugate | HRP |
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Product Type | Solid Phase Sandwich ELISA |
Catalog Number | Mfr. No. | Quantity | Sufficient For | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Sufficient For | Price | Quantity & Availability | ||||
MMA00
|
R&D Systems™
MMA00 |
2 Plates | Sufficient materials to run ELISAs on at least two 96-well plates |
Each for $922.20
|
|
||||
SMMA00
|
R&D Systems™
SMMA00 |
6 Plates | Sufficient materials to run ELISAs on at least six 96-well plates |
Each for $4,199.20
|
|
||||
Description
The Mouse CCL3/MIP-1 alpha Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse CCL3/MIP-1 alpha in cell culture supernates (50 ul), serum (50 ul).Specifications
4.70 - 300pg/mL | |
HRP | |
Solid Phase Sandwich ELISA | |
Natural and Recombinant Mouse MIP-1 alpha | |
ELISA | |
Cell Culture Supernates (50uL),Serum (50uL) | |
C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1 alpha, MIP1-alpha, PAT 464.1, SCYA3, SIS-beta |
1.5pg/mL | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
Cell Culture Supernatant, Serum | |
Mouse | |
20302 | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title