Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Naloxone hydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
500 mg
1 g
Description
A specific opiate antagonist. Blocks the action of sigma-agonists at opioid sites.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Naloxone hydrochloride the hydrochloride salt of the anti-opioid naloxone
Application
• Naloxone is a fast-acting inverse agonist of the mu-opioid receptor that can be used to competitively inhibit the binding of opioid compounds
Specifications
Melting Point | 200°C to 205°C |
Color | White |
Quantity | 100 mg |
Merck Index | 14,6362 |
Solubility Information | Soluble in water (73mg/ml),ethanol (3.3mg/ml),methanol (50mg/ml) and dimethyl sulfoxide (73mg/ml). Insoluble in ether |
Formula Weight | 363.84 |
Percent Purity | 98% |
Physical Form | Crystalline Powder |
Chemical Name or Material | Naloxone hydrochloride |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 206-611-0 |
RTECSNumber | QD2275000 |
TSCA | No |
storageNote1 | 2° to 8°C |
Recommended Storage | Keep cold |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction