Learn More
Neutec Group Smart Dilutor
For dilution of solid, semi-solid, or paste types of microbiological samples directly into sterile homogenizer bags
$8606.00 - $10606.00
Specifications
Product Type | Pipet Dispensing System |
---|---|
Autoclavable | Autoclavable |
Dilution | 0.1 g |
Volume (Metric) | 1 μL to 25 mL |
For Use With (Application) | Dilution of solid, semi solid, or paste types of microbiological samples directly into sterile homogenizer bags |
Includes: Both systems include polyethylene bag-holder, two sets of tubing, filters, 5L bottles, power cable, and an instruction manual. 14-910-315 is also supplied with two pumps.
Catalog Number | Mfr. No. | Model | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Includes | Price | Quantity & Availability | ||||
14-910-315
|
Neutec Group
90007900 |
Single pump | Polyethylene bag holder, two sets of tubing, filters, 5L bottles, power cable, instruction manual, one pump |
Each for $8,606.00
|
|
||||
14-910-316
|
Neutec Group
90007901 |
Two Pump | Polyethylene bag holder, two sets of tubing, filters, 5L bottles, power cable, instruction manual, two pump |
Each for $10,606.00
|
|
||||
Description
Since these types of samples require a gravimetric type of dilution process, the Smart Dilutor has an integrated balance at the heart of the system. The operator simply places an unknown amount of sample in a sterile bag and the unit takes an initial weight of the sample, then the operator moves the lever and the microprocessor dispenses an amount of diluent by weight, to achieve the programmed dilution ratio. The system constantly monitors the weight in the sample bag to achieve a highly accurate dilution result. Once the programmed dilution ratio has been reached, the system notifies the operator that the process has finished.
- Software also allows the instrument to be used as a dispensing device for a previously determined quantity of a liquid into any kind of container
- Bar code provides traceability of the dilution process
- Can be expanded to dilute up to six different buffers with the use of an optional accessory
- Adjustable, multilevel arm enables dilution of various type containers
- Tubing set and the bottles are fully autoclavable
- Changing operation of tubing set is performed in less than 15 seconds
Certifications
Each device is calibrated and certified against equipment whose calibration is traceable to NIST™
Specifications
Pipet Dispensing System | |
0.1 g | |
Dilution of solid, semi solid, or paste types of microbiological samples directly into sterile homogenizer bags | |
41°F to 104°F | |
7 x 6.13 x 13.13 in. (178 x 156 x 333 mm) | |
178 mm | |
156 mm | |
333 mm | |
6500 g | |
85/264 VAC | |
50/60 Hz |
Autoclavable | |
1 μL to 25 mL | |
950 mL/min. | |
5°C to 40°C | |
7 in. | |
6.13 in. | |
13.13 in. | |
5.2 lb. | |
85/264VAC 50/60Hz | |
40 w |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.