Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid (Certified ACS Plus), Fisher Chemical™

Catalog Number a200c212
Click to view available options
Quantity:
500 mL
2.5 L
6.5 gal.
4 x 2.5 L
Packaging:
Glass Bottle
Glass Carboy
Safe-Cote™ Glass
7697-37-2, 7732-18-5
HNO3
63.01
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
944
CHEBI:48107
nitric acid
O[N+]([O-])=O
7697-37-2, 7732-18-5
HNO3
63.01
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
944
CHEBI:48107
nitric acid
O[N+]([O-])=O

Nitric Acid material may darken during storage due to photochemical reaction

TRUSTED_SUSTAINABILITY
pH -1.0
Color Colorless Yellow
Melting Point -41°C
Boiling Point 120.5°C
Density 1.40g/cm³
Heavy Metals (as Pb) 0.2ppm max.
Identification Pass Test
Packaging Glass Bottle
Residue after Ignition 2ppm max.
Vapor Pressure 0.94 kPa at 20°C
Normality about 15.8
Assay Percent Range 70%
Quantity 4 x 2.5 L
Physical Form Liquid
UN Number 2031
Formula Weight 63.01
Percent Purity 68.0 to 70.0% (w/w)
Grade Certified ACS Plus
Chemical Name or Material Nitric Acid
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. May cause pulmonary edema. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:4
Flammability:0
Instability:0
DOTInformation DOT Class 8, : Corrosive
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit