Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nitric Acid 67-70%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™
Click to view available options
Quantity:
250 mL
500 mL
1 L
2 L
Packaging:
FEP bottle
Description
Nitric Acid, HNO3, CAS Number-7697-37-2, acide nitrique, acidum nitricum, azotic acid, rfna, aqua fortis, nitryl hydroxide, salpetersaeure, nitrous fumes, nital, hydrogen nitrate, 500mL
- Ideal for analysis by ICP-MS and high-resolution ICP-MS.
- Evaluated for 65 metals at low-level ppt concentrations.
- Manufactured, purified, and packaged in a Class 1000 clean room.
- Packaged in specially manufactured and cleaned FEP fluoropolymer bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
- Three year shelf life from time of manufacture - available on COA.
Specifications
pH | 1.0 |
Color | Colorless Yellow |
Melting Point | -41°C |
Boiling Point | 120.5°C |
Density | 1.40 g/cm3 |
Packaging | FEP bottle |
Vapor Pressure | 0.94 kPa at 20°C |
Certificate of Analysis | Certificate of Lot Analysis is included with each bottle. |
Quantity | 2 L |
Physical Form | Liquid |
Show More |
Safety and Handling
DOTInformation | DOT Class 8: Corrosive |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction