Learn More
Raylabcon Low Nitrogen Weighing Papers
SDPSCI_ED
Sturdy and nonabsorbent.
$20.25 - $61.00
Specifications
Product Type Low Nitrogen Paper
For Use With (Application) For analytical balances
View More Specs
Products 3
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
S402871
SDPSCI_ED
View Documents
Raylabcon
20 60 5623/EMD
3 x 3 in. (76 x 76mm)
Pack of 500 for $20.25
Case of 12 PK for $208.00
Add to Cart
S402881
SDPSCI_ED
View Documents
Raylabcon
20 60 5624/EMD
4 x 4 in. (100 x 100mm)
Pack of 500 for $33.25
Case of 12 PK for $362.00
Edge
Add to Cart
S402891
SDPSCI_ED
View Documents
Raylabcon
20 60 5625/EMD
6 x 6 in. (152 x 152mm)
Pack of 500 for $61.00
Case of 12 PK for $731.00
Add to Cart
Description
Description
Sturdy and nonabsorbent.
  • Supports sample
  • Smooth texture avoids trapping of material
  • Will not absorb moisture
  • Sold in packs of 500
  • Packaged in 4 mil poly bag for extra protection
Specifications
Specifications
Low Nitrogen Paper
For analytical balances