Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SouthernBiotech™ Normal Donkey Serum
SouthernBiotech normal sera are useful as blocking reagents in a variety of immunoassays.
Supplier: SouthernBiotech™ 003001
Description
Processed to minimize hemolysis and 0.2 μm filteredSpecifications
Supplied with 0.1% NaN3 as a preservative | |
Normal Donkey Serum | |
10 mL | |
Blocking |
Store at 2-8°C | |
Donkey Serum | |
Supplied with 0.1% NaN3 as a preservative |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use only
Spot an opportunity for improvement?
Provide Content Correction