Learn More
Ortho-Clinical Diagnostics™ VITROS™ 350 System
Up to 300 results per hour
Supplier: Ortho-Clinical Diagnostics™ 6802153
Description
Key Features
- On board capacity for all 45 assays
- Sample types: serum, plasma, urine, CSF
- Sample volume: 5–11μL
- Sample capacity: 40 samples (4 trays of 10 samples)
- Time for single results: 2.5 to 8 minutes
- Primary tube sampling
- On board dilution
- Unique sample processing capabilities like disposable tips, bubble, and clot detection assure confidence in results
- Does not require deionized water, plumbing, or drains
- True random access testing
- Low-glare flat screen offers easy, touch-sensitive operation with enhanced ergonomics
- Wide, accessible on board menu delivers immediate results for STAT or routing testing and disease-focused panels
- Customized patient reports for physician office labs
- High reagent efficiency
- “Load and Go” reagent preparation, MicroSlide™ technology, and up to six months' calibration stability mean labor can be redeployed for value-added tasks
- VITROS™ technology reduces interference from hemolysis, lipemia, and bilirubin
- Low cost per reportable result
- Minimal carryover and interference
- Minimal instrument maintenance
- Minimal chemical or infectious wastes reduce biohazards
- Self-monitoring alerts operators to prompt action avoiding testing delays
- Small footprint requires less space and overhead expenses
Specifications
Plasma, Serum, Urine or CSF | |
Colorimetry, potentiometry | |
100, 120/200 to 240 V 47/63 Hz | |
100, 120/200 to 240 V |
Yes | |
3600 Tests | |
47/63 Hz |