Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Owl™ EC1000XL Programmable Power Supply
Save benchtop space with the stackable design of this programmable power supply.
Supplier: Thermo Scientific™ EC1000XL
This item is not returnable.
View return policy
Description
Save benchtop space with the stackable design of the Thermo Scientific™ Owl™ EC1000XL Programmable Power Supply. Ideal for DNA, RNA and protein electrophoresis, as well as electroblotting, the power supply has a power-off memory that retains settings after shut-down.
- Power supply is simple to use, multi-purpose and lightweight
- Power supply includes soft-touch key pad and four jacks
Specifications
500 mA | |
500 mA | |
0 to 99 hr. 59 min. | |
4 | |
12 in. | |
10.75 in. | |
5.25 in. | |
120 V 50/60 Hz | |
120 V |
Model EC1000XL; 120V 50/60Hz | |
Owl | |
Voltage, current or power | |
12 x 10.75 x 5.25 in. (31 x 27 x 13.3 cm) | |
31 cm | |
27 cm | |
13.3 cm | |
1000 V | |
50/60 Hz |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction