Learn More
Perfex™ Neoprene Quick-Dry Foam Squeegees
Flexible blades are specifically designed for use on uneven or irregular surfaces
Supplier:  Perfex™ 9724
Catalog No. 19-165-126
Add to Cart
Description
Description
Twin-blade floor squeegee made of neoprene rubber with a durable polymer channel
  • L-N-T connection accepts polymer handle that adjusts from 86.4 to 157.5cm (34 to 62 in.)
  • Resistant to water, oil, grease, detergents, sanitizers and solvents
  • Washes and rinses clean with warm water and dries quickly
  • Handle sold separately
Specifications
Specifications
Floor Squeegee
Designed for use on uneven or irregular surfaces
Neoprene Rubber with a durable Polymer channel
Product Suggestions
Product Suggestions