Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphoric Acid - Triethylamine 1:1 Solution, Honeywell™ Fluka™
Click to view available options
Quantity:
100 mL
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Packaging | Glass Bottle |
Quantity | 100 mL |
Molecular Formula | C6H18NO4P |
Synonym | triethylamine phosphate, triethylammonium phosphate, triethylamine phosphate 1:1, n,n-diethylethanamine; phosphoric acid, phosphoric acid; triethylamine, ethanamine, n,n-diethyl-, phosphate, ethanamine, n,n-diethyl-, phosphate 1:?, ethanamine, n,n-diethyl-, phosphate 1:1, teap, acmc-20aj7n |
InChI Key | UNXNGGMLCSMSLH-UHFFFAOYSA-N |
SMILES | OP(O)(O)=O.CCN(CC)CC |
IUPAC Name | phosphoric acid; triethylamine |
Molecular Weight (g/mol) | 199.19 |
PubChem CID | 61946 |
Formula Weight | 199.19g/mol |
Safety and Handling
Hazard Statement | P261-P305 + P351 + P338 |
Precautionary Statement | H315-H319-H335 |
Recommended Storage | Store at 2° to 8°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction