Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Water, LC-MS Grade

Thermo Scientific™ Water, LC/MS Grade is ultapure water for formulation of solvents for HPLC

$97.00

Specifications

Color Clear, Colorless
Melting Point 32°F
Boiling Point 212°F
Grade LC-MS
Physical Form Liquid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PI51140
View Documents
Thermo Scientific™
T51140K2
1 L
Each for $97.00
Add to Cart
 
PI85189
View Documents
Thermo Scientific™
85189
4 x 1 L Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.

View alternate product
N/A
Description

Description

Thermo Scientific™ Water, LC/MS Grade is ultapure water for formulation of solvents for HPLC.

An ultrapure, LC/MS-grade preparation with low UV-absorptivity that makes it suitable and reliable for use in HPLC and mass spectrometry applications.

Thermo Scientific Water, LC/MS Grade is specially purified by a proprietary method and treated to ensure lot-to-lot consistency with a low UV absorbance to provide you with most sensitive detection across all wavelengths. Water is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen atmoshere with Teflon TFE-lined fluorocarbon caps for ultimate protection.

Highlights:

  • Purity and quality validated with 30 tests for use in LC/MS applications
  • 0.2-micron filtered, packed in solvent-rinsed amber glass bottles
  • Sealed under nitrogen atmosphere to eliminate absorption of unknown atmospheric gases

Product Details:

General properties of water

  • Molecular formula: H2O
  • Molecular Weight: 18.01
  • CAS number: 7732-18-5

Water, LC/MS Grade is specifically purified and tested to the highest specifications to ensure the integrity of your data, maximize sensitivity in your assay and prolong the life of your equipment.

Specifications

Specifications

Clear, Colorless
212°F
Liquid
32°F
LC-MS
Pierce™
Safety and Handling

Safety and Handling

Recommended Storage : Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit