Learn More
Corning™ PYREX™ Crystallizing Dishes
Reusable borosilicate glass dishes with heavy duty rims ideal for storage and crystallization
$219.00 - $491.00
Specifications
Autoclavable Autoclavable
Bottom Shape Flat
Disposable Reusable
Material Borosilicate Glass
Shape Round
View More Specs
Products 8
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Height (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Height (Metric) Price Quantity & Availability  
08-741B
View Documents
Corning™
314080
180 mL 80 mm 40 mm
Pack of 6 for $219.00
Case of 4 PK for $641.00
Edge
Add to Cart
08-741D
View Documents
Corning™
3140100
325 mL 100 mm 50 mm
Pack of 6 for $294.00
Case of 3 PK for $647.00
Add to Cart
08-741A
View Documents
Corning™
314070
180 mL 70 mm 50 mm
Pack of 6 for $226.00
Case of 4 PK for $658.00
Edge
Add to Cart
08-741C
View Documents
Corning™
314090
270 mL 90 mm 50 mm
Pack of 6 for $241.00
Case of 3 PK for $527.00
Add to Cart
08-741E
View Documents
Corning™
3140125
740 mL 125 mm 65 mm
Pack of 4 for $371.00
Case of 3 PK for $814.00
Edge
Add to Cart
08-741F
View Documents
Corning™
3140150
1200 mL 150 mm 75 mm
Pack of 4 for $491.00
Case of 2 PK for $718.00
Add to Cart
08-741G
View Documents
Corning™
3140170
1770 mL 170 mm 90 mm
Pack of 2 for $296.00
Case of 4 PK for $866.00
Add to Cart
08-741H
View Documents
Corning™
3140190
2500 mL 190 mm 100 mm
Pack of 2 for $350.00
Case of 3 PK for $768.00
Add to Cart
Description
Description
Reusable borosilicate glass dishes with heavy duty rims ideal for storage and crystallization
The rims on these dishes are reinforced and fire polished to reduce chipping. Will withstand repeated sterilization (wet or dry). Ideal for storage and crystallization.
Specifications
Specifications
Autoclavable
Reusable
Round
Ideal for storage and crystallization
Flat
Borosilicate Glass
Crystallizing Dish