Learn More
Qorpak™ SturdeeSeal™ Closure Torque System
Lightweight, portable and easy to use and store
Supplier: Qorpak™ HMS10150
This item is not returnable.
View return policy
Description
- Features two sets of interchangeable rubber inserts to accommodate 0.8 to 2.8 in. (20 to 70mm) closure sizes
- Closure adapter made of Delrin™ with aluminum frame and textured stainless-steel adjuster
- Secures closures at correct degree of torque to ensure proper seal
- For use with smooth finish and ribbed closures
Specifications
Glass and plastic containers |