Learn More
Quidel QuickVue™ hCG Urine Test
A single-step procedure
Supplier: Quidel 20109
Description
- CLIA Complexity: Waived
- Sensitive immunoassay for the qualitative detection of human chorionic gonadotropin (hCG) in urine
- Single-step technology uses a monoclonal antibody specific to the beta subunit of hCG to detect hCG
- Elevated hCG levels (>25mIU/mL) develop a pink-to-red test line and a blue procedural control line in the result window; very low levels of hCG (0 to 25mIU/mL) develop only a blue procedural control line in the result window
- For in vitro diagnostic use only
Specifications
Waived | |
Urine | |
25 Tests |
hCG | |
hCG Urine Test |