Learn More
R&D Systems™ CometAssay Single Cell Gel Electrophoresis Assay-Silver
Provides a simple and effective method for evaluating DNA damage in cells
$373.00 - $500.00
Specifications
Quantity | 50 Tests |
---|---|
Product Type | Single Cell Gel Electrophoresis Assay |
Includes: Lysis Solution, 2 x 500mL; Comet LMAgarose, 15mL; Trevigen CometSlide, 25/kit; pH 10 200mM EDTA, 12.5mL; Kit with Silver Stain (Mfr. No. 4251-050-K) also includes four staining reagents and a fixation additive.
Catalog Number | Mfr. No. | Product Type | Kit Contents | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Kit Contents | Price | Quantity & Availability | ||||
4251050K
|
R&D Systems™
4251050K |
Single Cell Gel Electrophoresis Assay | Lysis Solution, Comet LMAgarose, CometSlide, pH 10 200mM EDTA, four staining reagents and fixation additive, Silver Stain |
Each for $500.00
|
|
||||
4250050K
|
R&D Systems™
4250050K |
Single Cell Gel Electrophoresis Assay | Lysis Solution (2 x 500mL), Comet LMAgarose (15mL); Trevigen CometSlide (25/kit) and pH 10 200mM EDTA (12.5mL) |
Each for $373.00
|
|
||||
Description
The principle of the assay is based upon the ability of denatured, cleaved DNA fragments to migrate out of the nucleoid under the influence of an electric field, whereas undamaged DNA migrates slower and remains within the confines of the nucleoid when a
Choose the Neutral CometAssay to detect double-stranded breaks or the Alkaline CometAssay for a more sensitive protocol used to detect smaller amounts of damage including single and double-stranded breaks
Cells are immobilized in a bed of low melting poi
Specifications
50 Tests | |
Single Cell Gel Electrophoresis Assay |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.