Learn More
ZING™ Enterprises SDS Binder
Each binder contains a heavy-duty ring assembly, available in 3 sizes.
$42.60 - $60.70
Specifications
Description | Floor standing |
---|---|
Product Type | Hazard Communication |
Quantity | 1 |
Material | Plastic |
For Use With (Application) | SDS Management |
Catalog Number | Mfr. No. | Height (English) | Width (English) | Diameter (English) Ring | Languages | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Height (English) | Width (English) | Diameter (English) Ring | Languages | Price | Quantity & Availability | ||
18-003-013
|
ZING™ Enterprises
6030 |
12 in. | 1.5 in. | 1.5 in. | English |
Each for $42.60
|
|
||
18-003-014
|
ZING™ Enterprises
6031 |
12 in. | 2.5 in. | 2.5 in. | English |
Each for $54.10
|
|
||
18-003-015
|
ZING™ Enterprises
6032 |
12.5 in. | 3 in. | 3 in. | English |
Each for $60.70
|
|
||
18-003-016
|
ZING™ Enterprises
6033 |
12 in. | 1.5 in. | 1.5 in. | English, Spanish |
Each for $42.60
|
|
||
18-003-017
|
ZING™ Enterprises
6034 |
12 in. | 2.5 in. | 2.5 in. | English, Spanish |
Each for $54.10
|
|
||
18-003-018
|
ZING™ Enterprises
6035 |
12.5 in. | 3 in. | 3 in. | English, Spanish |
Each for $60.70
|
|
||
Description
ZING SDS Binder prominently stores important safety and training documents to comply with the new GHS/Hazard Communication requirements. Binder is constructed from high quality 100% post-consumer poly materials that reinforce sustainability initiatives as well as compliance. Binder is highly visible with its bright red/white colors and is extremely durable, chemical resistant and easy to clean.
Different sizes include – 1.5 in. (up to 300 sheets), 2.5 in. (up to 500 sheets), and 3 in. (up to 600 sheets). Binders are available in English and bilingual English/Spanish versions.
Specifications
Floor standing | |
1 | |
SDS Management |
Hazard Communication | |
Plastic |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.