Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Shandon™ Artery Scissors, Straight, Sharp/Sharp, standard, 4.5 in. (11.4cm)
Cut with precision using Epredia™ Shandon™ Artery Scissors, available in premium and standard sizes.
Supplier: Epredia™ 3120007
Description
Consistent Quality
- Properly hardened, tempered and passivated
- Electronically polished and cleaned for a high-quality finish
- Patterns, dimensions and finish held to exacting standards
- Guaranteed to be free from defects in material and workmanship
Specifications
Artery Scissors | |
Standard | |
11.4 cm |
Straight | |
4.5 in. | |
Sharp |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction