Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ SIS3
Selective Smad3 inhibitor; inhibits TGF-βR1 signaling
$388.60 - $1599.64
Specifications
Purity | 97% |
---|---|
Description | SIS3 |
Molecular Weight (g/mol) | 489.99 |
Product Type | SIS3 |
Description
Selective Smad3 inhibitor; attenuates TGF-β1-dependent Smad3 phosphorylation and DNA binding. Has no effect on Smad2, p38 MAPK, ERK or PI 3-kinase signaling. Inhibits TGF-β1-induced myofibroblast differentiation of dermal fibroblasts. Also inhibits TGF-β2-induced endothelial cell differentiation in iPSCs.Specifications
97% | |
SIS3 | |
489.99 | |
SIS3 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title