Learn More
ICU Medical Jelco™ Saf-T Holder™ Blood Culture Holders and Transfer Devices
Saf-T Holder™ devices come with the back-end needle and multi-sample Luer permanently attached to the holder to minimize potential exposures and holder re-use. Available separately or with Saf-T Wing™ Blood Collection & Infusion Sets, Saf-T Holder™ devices also provide easy needless transfer after a syringe draw.
Specifications
For Use With (Application) | Multi-sample Luer permanently attached to the holder, provide easy needless transfer after a syringe draw |
---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
22-024-900
|
ICU Medical
96004 |
Saf-T Holder™ Blood Culture Device, with male luer |
Case of 50 for $211.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-024-901
|
ICU Medical
96005 |
Saf-T Holder™ Blood Culture Device, with female luer |
Case of 50 for $211.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- Eliminate potential exposures and holder reuse
- Provide safe needleless transfer after a syringe draw
Specifications
Multi-sample Luer permanently attached to the holder, provide easy needless transfer after a syringe draw |