Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ MediaKap™ and MediaKap Plus HF Filters
SDP

0.2μm DynaFibre™ mixed cellulose ester filter provides more surface area for faster flow than disc filters with less fouling

Specifications

Material Polycarbonate, Polyurethane
Material (Membrane) MCE
Pore Size 0.2 μm
Sterility Sterile
For Use With (Application) Efficiently sterilize and clarify culture medium or buffer solution
View More Specs

Products 11
Catalog Number Mfr. No. Connectors Filter Type Includes Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Connectors Filter Type Includes Volume (Metric) Price Quantity & Availability  
08-671-2B
SDP
View Documents
Spectrum™
ME2M02B12S
Female luer lok/0.25HB MediaKap-2 Automatic vent to prevent air lock, filling bell 0.2 to 2 L Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
08-607-033
SDP
View Documents
Spectrum™
ME2M02018S
Female luer lok/0.25HB MediaKap-2 Automatic vent to prevent air lock 0.2 to 2 L Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area. N/A
08-671-2E
SDP
View Documents
Spectrum™
ME2M10018S
0.25HB/0.25HB MediaKap-10 Automatic vent to prevent air lock 5 to 10 L Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
08-671-2F
SDP
View Documents
Spectrum™
ME2M10B12S
0.25HB/0.25HB MediaKap-10 Automatic vent to prevent air lock, filling bell 5 to 10 L Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area. N/A
08-671-2G
SDP
View Documents
Spectrum™
ME2M25B06S
VarHB/VarHB MediaKap-25 Automatic vent to prevent air lock, filling bell 10 to 25 L Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area. N/A
08-671-2H
SDP
View Documents
Spectrum™
ME2M50B03S
VarHB/VarHB MediaKap-50 Automatic vent to prevent air lock, filling bell 25 to 50 L Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area. N/A
08-607-039
SDP
View Documents
Spectrum™
MP2M50B03S
VarHB/VarHB MediaKap-50 Plus Automatic vent to prevent air lock, filling bell 25 to 50 L Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
08-671-4C
SDP
View Documents
Spectrum™
MP2M05018S
Female luer lok/0.25HB MediaKap-5 Plus Automatic vent to prevent air lock 2 to 5 L Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
08-671-4F
SDP
View Documents
Spectrum™
MP2M25B06S
VarHB/VarHB MediaKap-25 Plus Automatic vent to prevent air lock, filling bell 10 to 25 L Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
08-671-4E
SDP
View Documents
Spectrum™
MP2M10B12S
0.25HB/0.25HB MediaKap-10 Plus Automatic vent to prevent air lock, filling bell 5 to 10 L Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
08-671-4D
SDP
View Documents
Spectrum™
MP2M05B12S
Female luer lok/0.25HB MediaKap-5 Plus Automatic vent to prevent air lock, filling bell 2 to 5 L Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

  • Efficiently sterilize and clarify culture medium or buffer solution
  • Smaller, narrow design for low hold-up volume
  • Polypropylene vent prevents air lock
  • Operates with peristaltic pump, vacuum, or syringe
  • Available in five different sizes that filter designated volume in 15 to 20 minutes
  • Sterile, biocompatible and nonpyrogenic
  • USP Class VI; 100% integrity tested

  • Optional filling bell maintains sterility while filling bottles
  • MediaKap Plus filters utilize an advanced DynaFibre membrane that further increases filtration efficiency and dramatically reduces the time required to filter serum-enriched medium

Specifications

Specifications

Polycarbonate, Polyurethane
0.2 μm
Efficiently sterilize and clarify culture medium or buffer solution
MCE
Sterile
Bottle Top Filter
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit