Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENTiCARE Stainless Steel IV Pole with Five Legged Deluxe Base and Eight Hooked Rake

Features hook rake to provide easy management of multiple IV lines

Supplier:  CENTiCARE IV1000D8

Encompass

Catalog No. 22-555-235


Add to Cart
This item is not returnable. View return policy

Description

Description

  • Five-legged polished aluminum base (Diameter: 543.36cm)
  • Eight rake top for line management
  • Oversized adjustment knob
  • Wheels with Carpet Master™ deluxe casters (Diameter: 7.62cm)

Service Plan

No

Warranty and Services

One year

Specifications

Specifications

IV Poles
Eight hook rake
Stainless Steel
With silent casters
Transportation of patient supplies in hospitals
90.06 kg
5
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit