Learn More
Thermo Scientific™ SureTect™ Real-Time PCR System
Quickly and accurately detect microorganisms in a broad range of food and environmental samples using the Thermo Scientific™ SureTect™ System.
$17.00 - $4898.00
Specifications
For Use With (Application) SureTect Real Time PCR Assays
View More Specs
Products 4
Catalog Number Mfr. No. Description For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. Description For Use With (Application) Price Quantity & Availability  
PT0500W
Accessories View Documents
Thermo Scientific™
PT0500W
SureTect Software SureTect Real Time PCR Assays
Each for $17.00
Add to Cart
PT0800
Accessories View Documents
Thermo Scientific™
PT0800
SureTect Laptop Computer SureTect Real Time PCR Assays
Each for $4,898.00
Add to Cart
PT0600
Accessories View Documents
Thermo Scientific™
PT0600
SureTect Start-up Package SureTect Real Time PCR Assays Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
PT0700
Accessories View Documents
Thermo Scientific™
PT0700
SureTect Desktop Computer SureTect Real Time PCR Assays Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description
Description
Quickly and accurately detect microorganisms in a broad range of food and environmental samples using the Thermo Scientific™ SureTect™ System. The system combines a real-time PCR instrument with bespoke software and convenient pre-filled reagent components that, together with a single PCR protocol for a range of common targets and sample types, streamline the test workflow.
The SureTect System is a unique solution combining speed and performance in an easy-to-use, cost-effective platform—giving you results you can trust.
  • Plug and Play:No installation specialist required
  • Streamlined Protocol: Facilitates processing of multiple assays in the same run, for maximum efficiency
  • Simple, Intuitive Software: Enables rapid, straightforward PCR set-up, automatic data interpretation and traceability of results
  • Small Instrument Footprint: Fits easily into laboratory environments, where bench space is at a premium
  • LED Light Technology: Long lasting, meaning no dimming of light over time
  • USB Port: Enables system to be run without a laptop
Specifications
Specifications
SureTect Real Time PCR Assays