Learn More
Synthware™ Pressure Equalizing Funnels with High Vacuum Valve
With extended tip valve for semi-needle valve adjustment
$209.50 - $291.00
Specifications
Diameter (Metric)Top | Valve orifice: 0-4mm |
---|---|
Product Type | Pressure Equalizing Funnel with High Vacuum Valve |
Catalog Number | Mfr. No. | Capacity (Metric) | Joint Number | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Joint Number | Price | Quantity & Availability | ||||
31-501-725
|
Synthware™
F641425 |
25 mL | 14/20 |
Each for $209.50
|
|
||||
31-501-726
|
Synthware™
F641460 |
60 mL | 14/20 |
Each for $229.50
|
|
||||
31-501-727
|
Synthware™
F641960 |
60 mL | 19/22 |
Each for $229.50
|
|
||||
31-501-728
|
Synthware™
F642460 |
60 mL | 24/40 |
Each for $280.00
|
|
||||
31-501-729
|
Synthware™
F641125 |
125 mL | 14/20 |
Each for $236.50
|
|
||||
31-501-730
|
Synthware™
F649125 |
125 mL | 19/22 |
Each for $229.50
|
|
||||
31-501-731
|
Synthware™
F644125 |
125 mL | 24/40 |
Each for $273.00
|
|
||||
31-501-732
|
Synthware™
F644250 |
250 mL | 24/40 |
Each for $291.00
|
|
||||
Description
- With pressure equalizing arm and high vacuum valve
- Top outer and lower inner standard taper joints of same size
Specifications
Valve orifice: 0-4mm | |
Pressure Equalizing Funnel with High Vacuum Valve |