Learn More
Synthware™ Addition Funnels with Pressure Equalizing Arm and PTFE Stopcock
Cylindrical style addition funnel with pressure equalizing arm and PTFE Stopcock
$290.50 - $439.00
Catalog Number | Mfr. No. | Capacity (Metric) | Joint Number | Description | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Joint Number | Description | Price | Quantity & Availability | |||
31-501-719
|
Synthware™
F671425 |
25 mL | 14/20 | Stopcock bore: 2mm |
Each for $290.50
|
|
|||
31-501-720
|
Synthware™
F672460 |
60 mL | 24/40 | Stopcock bore: 2mm |
Each for $324.00
|
|
|||
31-501-721
|
Synthware™
F674125 |
125 mL | 24/40 | Stopcock bore: 2mm |
Each for $333.00
|
|
|||
31-501-722
|
Synthware™
F674250 |
250 mL | 24/40 | Stopcock bore: 2mm |
Each for $363.00
|
|
|||
31-501-723
|
Synthware™
F674500 |
500 mL | 24/40 | Stopcock bore: 4mm |
Each for $377.00
|
|
|||
31-501-724
|
Synthware™
F67241L |
1000 mL | 24/40 | Stopcock bore: 4mm |
Each for $439.00
|
|
|||
Description
- Top outer and lower inner standard taper joints of same size
Specifications
Pressure Equalizing Funnel with PTFE stopcock |