Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Total Ionic Strength Adjustment Buffer (TISAB II), with CDTA, Ricca Chemical
For Fluoride Analysis using Ion Selective Electrodes
$146.30 - $1334.00
Specifications
Chemical Name or Material | Buffer Solution, Total Ionic Strength Adjustment Buffer (TISAB II) |
---|---|
Name Note | With CDTA |
CAS Min % | 82.0, 5.87, 4.91, 4.77, 0.33 |
CAS Max % | 85.0, 6.11, 5.12, 4.96, 0.34 |
CAS | 1310-73-2, 64-19-7, 7647-14-5, 13291-61-7 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
8670-16
|
Ricca Chemical Company
867016 |
500 mL | Natural Poly Bottle |
Each for $146.30
|
|
||||
8670-32
|
Ricca Chemical Company
867032 |
1 L | Natural Poly Bottle |
Each for $182.60
|
|
||||
8670-1
|
Ricca Chemical Company
86701 |
4 L | Natural Poly Bottle |
Each for $311.50
|
|
||||
867025
|
Ricca Chemical Company
86702.5 |
10 L | Cubitainer |
Each for $835.00
|
|
||||
8670-5
|
Ricca Chemical Company
86705 |
20 L | Cubitainer |
Each for $1,334.00
|
|
||||
Specifications
Buffer Solution, Total Ionic Strength Adjustment Buffer (TISAB II) | |
82.0, 5.87, 4.91, 4.77, 0.33 | |
1310-73-2, 64-19-7, 7647-14-5, 13291-61-7 |
With CDTA | |
85.0, 6.11, 5.12, 4.96, 0.34 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title