Learn More
Troemner™ Individual Analytical Precision Weights, Class 1 with NVLAP Certificate
Low magnetic susceptibility, uniform consistency and high resistance to corrosion
$278.00 - $2760.00
Specifications
Certifications/Compliance | NVLAP, ISO9001 |
---|---|
Classification | ASTM Class 1 |
Includes | Polycarbonate case |
Quantity | Each |
Packaging Quantity | 1 |
Catalog Number | Mfr. No. | Product Type | Mass | Shape | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Mass | Shape | Price | Quantity & Availability | |||
02-214-1JW
|
Troemner™
80781144 |
Two-Piece Metric Analytical Weight | 200 g | Cylindrical with Handling Knob |
Each for $598.50
|
|
|||
02-319-1AC
|
Troemner™
80781142 |
Two-Piece Metric Analytical Weight | 100 g | Cylindrical with Handling Knob |
Each for $376.50
|
|
|||
02-319-1BC
|
Troemner™
80781140 |
Two-Piece Metric Analytical Weight | 50 g | Cylindrical with Handling Knob |
Each for $363.00
|
|
|||
02-319-1CC
|
Troemner™
80781136 |
Two-Piece Metric Analytical Weight | 20 g | Cylindrical with Handling Knob |
Each for $356.00
|
|
|||
02-319-1DC
|
Troemner™
80781134 |
Two-Piece Metric Analytical Weight | 10 g | Cylindrical with Handling Knob |
Each for $342.50
|
|
|||
02-319-1EC
|
Troemner™
80781132 |
Two-Piece Metric Analytical Weight | 5 g | Cylindrical with Handling Knob |
Each for $342.50
|
|
|||
02-319-1FC
|
Troemner™
80781128 |
Two-Piece Metric Analytical Weight | 2 g | Cylindrical with Handling Knob |
Each for $311.50
|
|
|||
02-319-1GC
|
Troemner™
80781126 |
Two-Piece Metric Analytical Weight | 1 g | Cylindrical with Handling Knob |
Each for $289.50
|
|
|||
02-319-1HC
|
Troemner™
80781124 |
One Piece Metric Analytical Sheet Weight | 500 mg | Flat with one end turned up for easy handling |
Each for $278.00
|
|
|||
02-319-1JC
|
Troemner™
80781120 |
One Piece Metric Analytical Sheet Weight | 200 mg | Flat with one end turned up for easy handling |
Each for $278.00
|
|
|||
02-319-1KC
|
Troemner™
80781118 |
One Piece Metric Analytical Sheet Weight | 100 mg | Flat with one end turned up for easy handling |
Each for $278.00
|
|
|||
02-319-1PC
|
Troemner™
80781110 |
One Piece Metric Analytical Sheet Weight | 10 mg | Flat with one end turned up for easy handling |
Each for $278.00
|
|
|||
02-319-1QC
|
Troemner™
80781108 |
One Piece Metric Analytical Sheet Weight | 5 mg | Flat with one end turned up for easy handling |
Each for $278.00
|
|
|||
02-319-1RC
|
Troemner™
80781104 |
One Piece Metric Analytical Sheet Weight | 2 mg | Flat with one end turned up for easy handling |
Each for $278.00
|
|
|||
02-319-1TC
|
Troemner™
80781102 |
One Piece Metric Analytical Sheet Weight | 1 mg | Flat with one end turned up for easy handling |
Each for $278.00
|
|
|||
02-214-118
|
Troemner™
30391440 |
Two-Piece Metric Analytical Weight | 3 g | Cylindrical with Handling Knob |
Each for $311.50
|
|
|||
02-216-001
|
Troemner™
30391453 |
Two-Piece Metric Analytical Weight | 5 kg | Cylindrical with Handling Knob |
Each for $2,760.00
|
|
|||
02-100-142
|
Troemner™
80781116 |
One Piece Metric Analytical Sheet Weight | 50 mg | Flat with one end turned up for easy handling |
Each for $278.00
|
|
|||
Description
Designed so the specifications on surface finish, markings, construction, material density and magnetic properties get more restrictive as the weight tolerances get smaller in order to enable the weight to maintain its tolerance over time
- Used in extremely stable environments
- 1mg to 500mg are 1 piece construction with no adjusting cavity with one end turned up for easy handling
- 1g to 5kg are 2 piece construction with handling knob
- Surfaces are polished to a perfect, mirror-like finish
Can be used as a reference standard in calibrating other weights and is appropriate for calibrating high-precision analytical balances with a readability as low as 0.1mg to 0.01mg
Specifications
NVLAP, ISO9001 | |
Polycarbonate case | |
1 |
ASTM Class 1 | |
Each |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.