Learn More
Thermo Scientific™ Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers
Secure vessels using these universal platform clamps for orbital shakers.
$53.90 - $1241.00
Catalog Number | Mfr. No. | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
11-676-011
|
Thermo Scientific™
236010 |
25mL Erlenmeyer Flask Clamp | MaxQ™ 8000 Shaker |
Each for $53.90
|
|
||||
11-676-012
|
Thermo Scientific™
236011 |
50mL Erlenmeyer Flask Clamp | MaxQ™ 8000 Shaker |
Each for $62.50
|
|
||||
11-676-013
|
Thermo Scientific™
236012 |
125mL Erlenmeyer Flask Clamp | MaxQ™ 8000 Shaker |
Each for $65.90
|
|
||||
11-676-014
|
Thermo Scientific™
236013 |
250/300mL Erlenmeyer Flask Clamp | MaxQ™ 8000 Shaker |
Each for $69.60
|
|
||||
11-676-015
|
Thermo Scientific™
236014 |
500mL Erlenmeyer Flask Clamp | MaxQ™ 8000 Shaker |
Each for $118.80
|
|
||||
11-676-016
|
Thermo Scientific™
236015 |
1L Erlenmeyer Flask Clamp | MaxQ™ 8000 Shaker |
Each for $141.90
|
|
||||
11-676-017
|
Thermo Scientific™
236016 |
2L Erlenmeyer Flask Clamp | MaxQ™ 8000 Shaker |
Each for $173.50
|
|
||||
11-676-018
|
Thermo Scientific™
236019 |
2800mL Fernbach Flask Clamp | MaxQ™ 8000 Shaker |
Each for $291.00
|
|
||||
11-676-019
|
Thermo Scientific™
236025 |
Microplate Clamp | MaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000 |
Each for $72.80
|
|
||||
11-676-002
|
Thermo Scientific™
161033 |
Vial/Tube Inserts | MaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000 |
Each for $347.00
|
|
||||
236017
|
Thermo Scientific™
236017 |
4L Erlenmeyer Flask Clamp | MaxQ HP 416, 420, 430, 435, 436, 480, 481 |
Each for $250.50
|
|
||||
236018
|
Thermo Scientific™
236018 |
6L Erlenmeyer Flask Clamp | MaxQ HP 416, 420, 430, 435, 436, 480, 481 |
Each for $230.00
|
|
||||
236028
|
Thermo Scientific™
236028 |
5L Erlenmeyer Flask Clamp | MaxQ HP 416, 420, 430, 435, 436, 480, 481 |
Each for $239.00
|
|
||||
11-676-020
|
Thermo Scientific™
236099 |
Flask Clamp Starter Kit for MaxQ High Performance Shakers | MaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000 |
Each for $1,241.00
|
|
||||
Description
Secure vessels using Thermo Scientific™ Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers. With exceptional innovation, reliability and accuracy, these orbital shakers, platforms and accessories are the optimal choice for a busy lab.
Flask Clamp Starter Kit includes:- (1) microplate clamp
- (2) 25mL flask clamps
- (2) 50mL flask clamps
- (2) 125mL flask clamps
- (4) 250/300mL flask clamps
- (4) 500mL flask clamps
- (2) 1L flask clamps
- (2) 2L flask clamps
- Replacement screws
Note:
Replacement screws also available separately.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.