Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Justrite™ VaporTrap™ HDPE, UN/DOT-Certified Carboy with Filter Kit, Four Ports 1/8 in., Three Ports 1/4 in., and One Port 1/4 in. Or 3/8 in. Hose Barb
Click to view available options
Capacity (Metric):
13.5 L
20 L
Description
- For packaging group II and hazardous chemicals
- Carbon filter prevents volatile organic compound vapors
- Two-piece cap design
Specifications
Product Type | UN/DOT Carboy with Filter Kit |
Capacity (English) | 5.3 gal. |
Certifications/Compliance | UN/DOT |
Closure Material | Polypropylene |
Closure Size | 70 mm |
Description | Four (4) 1/8 in. Tubing OD Ports, Three (3) 1/4 in. Tubing OD Ports and One (1) 1/4 or 3/8 in. Tubing ID Hose Barb Ports |
Capacity (Metric) | 20 L |
Dimensions (L x W x H) | 287 x 235 x 445 mm |
Material | HDPE |
For Use With (Application) | Safe transport of hazardous liquids |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction