Learn More
Vileda Professional™ CleanTech™ Duo Microfiber Laminated Mop, Sterile
For cleaning/disinfecting floors, walls and ceilings. Vileda Professional CleanTech Duo Microfiber Laminated Mop, Sterile are designed for use in cleanrooms up to class ISO 5/GMP A and B/FED209D Class 100.
Supplier: Vileda Professional™ 147544
Description
- Microfiber laminated cleanroom foam
- Double sided for effective cleaning and disinfecting
- Adhesive-free lamination techniques used to prevent additional contamination to products
- Suitable for bucket and pre-prepared method
- Proved bacteria reduction of 99.99%
- Validated to the 10-6 SAL per ISO 11137
- Individually double bagged with Lot # and Indicator Dot on each package
- Certificate of Sterility available upon request