Learn More
Thermo Scientific™ Waterproof Cryo Gloves - Mid-Arm Length
Catalog Number 13262785
Click to view available options
Size:
Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Safely handle samples from ultra low temperature freezers and cryogenic storage with Thermo Scientific™ Waterproof Cryo Gloves. Available in elbow, mid-arm, and shoulder-length options.
  • Provides splash protection
  • Comfortable, lightweight, breathable
  • Side release buckles for quick on/off
TRUSTED_SUSTAINABILITY
Description Mid-Arm Length
Product Type Waterproof Cryogenic Gloves
Size Large
For Use With (Application) Cryogenic Preservation
Length (English) 14 to 15 in.
Length (Metric) 35.5 to 38 cm