Learn More
Cytiva Whatman™ Mini-UniPrep™ Syringeless Filters
Processes sample loads in one-third the time. Whatman™ Mini-UniPrep™ Syringeless Filters provide a faster and easier way to remove particulates from samples being prepared for HPLC and UHPLC analysis.
$724.00 - $5860.00
Specifications
For Use With (Application) Routine HPLC/UHPLC analysis; Sample filtration; Composite assays; Content uniformity; Protein precipitation; Solubility/dissolubility testing; Ion chromatography
Type Syringeless Filter
View More Specs
Products 19
Catalog Number Mfr. No. Material (Membrane) Pore Size No. per Pack Price Quantity & Availability  
Catalog Number Mfr. No. Material (Membrane) Pore Size No. per Pack Price Quantity & Availability  
09-923-25
View Documents
Cytiva
UN203NPUAQU
PVDF 0.45 μm 100/Pk.
Pack of 100 for $724.00
Add to Cart
09-923-26
View Documents
Cytiva
UN203NPUDPP
Polypropylene Depth Filter 0.45 μm 100/Pk.
Pack of 100 for $724.00
Add to Cart
09-923-30
View Documents
Cytiva
UN203NPUPES
PES 0.45 μm 100/Pk.
Pack of 100 for $750.00
Add to Cart
09-923-36
View Documents
Cytiva
UN503NPUNYL
Nylon 0.45 μm 1000/Pk.
Pack of 1000 for $5,650.00
Edge
Add to Cart
09-923-28
View Documents Promotion Details
Cytiva
UN203NPUORG
PTFE 0.45 μm 100/Pk.
Pack of 100 for $724.00
Edge
Add to Cart
09-924-587
View Documents
Cytiva
UN503NPUPES
PES 0.45 μm 1000/Pk.
Pack of 1000 for $5,860.00
Add to Cart
09-923-29
View Documents
Cytiva
UN203NPUNYL
Nylon 0.45 μm 100/Pk.
Pack of 100 for $724.00
Add to Cart
09-923-106
View Documents
Cytiva
UN503NPENYL
Nylon 0.2 μm 1000/Pk.
Pack of 1000 for $4,396.00
Add to Cart
09-924-586
View Documents
Cytiva
UN503NPUDPP
Polypropylene Depth Filter 0.45 μm 1000/Pk.
Pack of 1000 for $5,860.00
Add to Cart
09-923-101
View Documents
Cytiva
UN203NPENYL
Nylon 0.2 μm 100/Pk.
Pack of 100 for $724.00
Add to Cart
09-923-32
View Documents
Cytiva
UN503NPUAQU
PVDF 0.45 μm 1000/Pk.
Pack of 1000 for $4,750.00
Add to Cart
09-923-27
View Documents
Cytiva
UN203NPUGMF
Glass Microfiber 0.45 μm 100/Pk.
Pack of 100 for $724.00
Add to Cart
09-923-102
View Documents
Cytiva
UN203NPEORG
PTFE 0.2 μm 100/Pk.
Pack of 100 for $724.00
Add to Cart
09-923-34
View Documents
Cytiva
UN503NPUGMF
Glass Microfiber 0.45 μm 1000/Pk.
Pack of 1000 for $5,650.00
Add to Cart
09-923-105
View Documents
Cytiva
UN503NPEAQU
PVDF 0.2 μm 1000/Pk.
Pack of 1000 for $5,650.00
Add to Cart
09-923-100
View Documents
Cytiva
UN203NPEAQU
PVDF 0.2 μm 100/Pk.
Pack of 100 for $724.00
Add to Cart
09-927-651
View Documents
Cytiva
UN203NPEPES
PES 0.2 μm 100/Pk.
Pack of 100 for $750.00
Add to Cart
09-923-31
View Documents
Cytiva
UN203NPUPP
Polypropylene 0.45 μm 100/Pk. Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
09-923-38
View Documents
Cytiva
UN503NPUPP
Polypropylene 0.45 μm 1000/Pk. Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description
Description
Processes sample loads in one-third the time. Whatman™ Mini-UniPrep™ Syringeless Filters provide a faster and easier way to remove particulates from samples being prepared for HPLC and UHPLC analysis.
Whatman Mini-UniPrep™ Transparent Syringeless Filters quickly and easily remove particulates from samples being prepared for HPLC/UHPLC analysis, while maintaining sample visibility.
  • Pre-assembled all-in-one filtration unit eliminates the need for syringe filter, syringe, vial and septum, reducing costs by up to 40%
  • Wide range of membrane choices in 0.2 and 0.45 μm pore sizes to meet specific sample application requirements
  • Compatible with most major autosamplers for high-throughput analysis
  • Fewer consumables required
  • Plunger contains a filtration membrane at one end and a pre-attached cap/septum at the other
  • Can be used either manually or with a compressor unit
  • Multi-sample compressor processes up to 6 samples at a time, further improving sample processing time and reducing hand stress
  • Designed to fit into any autosampler accommodating 12 x 32mm vials -OR-
    Septum can be pierced with needle and sample drawn off for manual injection into an analyzer
Specifications
Specifications
Routine HPLC/UHPLC analysis; Sample filtration; Composite assays; Content uniformity; Protein precipitation; Solubility/dissolubility testing; Ion chromatography
Syringeless Filter