Learn More
DWK Life Sciences Wheaton™ Headspace Standard Seals
Pressure release seals feature score lines that allow for internal pressure release when 3.0 ± 0.5 bar has been exceeded
$89.20 - $122.40
Specifications
Closure Color | Natural |
---|---|
Closure Material | Aluminum |
Type | Headspace Crimp Seal |
For Use With (Equipment) | Crimp Top Headspace Vials |
Catalog Number | Mfr. No. | Septum | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Septum | Price | Quantity & Availability | |||||
03-410-724
|
DWK Life Sciences
W224221 |
PTFE, Silicone |
Case of 100 for $107.90
|
|
|||||
03-410-726
|
DWK Life Sciences
W224225 |
PTFE, Gray Butyl Molded |
Case of 100 for $122.40
|
|
|||||
03-410-725
|
DWK Life Sciences
W224224 |
PTFE, Butyl |
Case of 100 for $89.20
|
|
|||||
Description
- Seals are made from aluminum for easy crimping and decapping
Specifications
Natural | |
Aluminum | |
Headspace Crimp Seal | |
Crimp Top Headspace Vials |