Username:  
Password:
Store:
New User?    Forgot Password?
Home
Help
Part of Thermo Fisher Scientific 
- Immunology/Serology
Browse Results: 146 item(s)
You're Browsing:Immunology/Serology
  1. 1
  2. 2
  3. 3
  4. 4
  5. 5
  6. >
  7. >>
1 Fisher HealthCare Sure-Vue* SELECT Rubella Latex Test
Does not require a dilution step for 10 IU sensitivity CLIA Complexity: Non-Waived   ·  CPT Codes: 86762/86406  ·  Sensitivity: 100%  ·  Specificity: 98.6%  ·  Rapid latex agglutination test for the qualitative and semi-quantitative
2 Accessories for Fisher HealthCare Sure-Vue* Test Kits
  ·  Controls for Fisher Scientific* Sure-Vue* Urine hCG, Sure-Vue* Serum/Urine hCG, and Sure-Vue* hCG STAT Test Kits  ·   Urine Controls STAT Controls Serum Controls
3 Fisher HealthCare Sure-Vue* Color Staph ID Latex Test Kit
Accuracy rating of 99.5%   ·  CLIA Complexity: Non-Waived  ·  CPT Code: 87077  ·  Sensitivity: 99%   ·  Specificity: 100%  ·  Latex agglutination slide test for the qualitative detection
4 Fisher HealthCare Sure-Vue* Mono Latex Test Kit
Qualitative and semi-quantitative results   ·  CPT Code: 86308  ·  CLIA Complexity: Non-Waived  ·  Latex agglutination slide test designed for the qualitative and semi-qauntitative detection of infectious mononucleosis heterophile antibodies in serum and
5 Fisher HealthCare Sure-Vue* RPR Test Kit
Variation of the standard VDRL CLIA Complexity: Non-Waived CPT Codes: 86542/86593   ·  Cardiolipin charcoal microparticle test for the qualitative and semi-quantitative determination of reagin, which is associated with syphilis  ·  Antigen
6 Fisher HealthCare Sure-Vue* Color Mono Test Kit
Ideal for standardization   ·  CPT Code: 86308  ·  CLIA Complexity: Non-Waived   ·  Color-enhanced slide test designed for detection of infectious mononucleosis heterophile antibodies in serum or plasma  ·  Test results in two minutes   ·  Specificity:
7 Fisher HealthCare Sure-Vue* Rota Test Kit
Simple two-step procedure   ·  CLIA Complexity: Non-Waived  ·  CPT Code: 87425  ·  Rapid membrane-based immunogold assay for the qualitative detection of rotavirus antigen in human fecal specimens; aids in the diagnosis of acute gastroenteritis caused by
8 Fisher HealthCare Sure-Vue* ASO Test Kit
Increased sensitivity ideal for testing pediatric patients   ·  CPT Codes: 86060/86063  ·  CLIA Complexity: Non-Waived  ·  Two-minute latex agglutination slide test   ·  For the qualitative and semi-quantitative determination of antistreptolysin-O
9 Fisher HealthCare Sure-Vue* CRP Latex Test Kit
Features a suspension of latex particles coated with the IgG fraction of an anti-human, CRP-specific serum   ·  CPT Code: 86140/86318  ·  CLIA Complexity: Non-Waived  ·  Sensitivity: 6mg/L serum   ·  Latex agglutination slide test   ·  Test
10 Fisher HealthCare Sure-Vue* RF Rapid Latex Test Kit
Suspension of latex particles is coated with human immunoglobulin   ·  CPT Codes: 86430/86318  ·  Latex agglutination test for the qualitative and semiquantitative determination of rheumatoid factor in serum   ·  CLIA Complexity: Non-Waived  ·  Can
11 Fisher HealthCare Sure-Vue* Rubella Latex Test Kit
Requires a dilution step for 10 IU sensitivity CLIA Complexity: Non-Waived CPT Codes: 86762/86406   ·  Sensitivity: 99.5%   ·  Specificity: 96.1%  ·  Non-dilution: 1-2 IU sensitivity; dilution: 10 IU sensitivity  ·  Latex
12 Fisher HealthCare Sure-Vue* Strep A Lateral Flow Test Kit
One-step lateral-flow immunoassay for the qualitative detection of Group A Strep from throat swabs   ·  CLIA Complexity: Waived and Moderate   ·  CPT Code: 87081  ·  Sensitivity: 97%  ·  Specificity:
13 Fisher HealthCare Sure-Vue* Adeno Test Kit
Simple two-step procedure   ·  CPT Code: 87301  ·  CLIA Complexity: Moderate   ·  Results in 15 minutes  ·  Rapid, membrane-based immunogold assay for the qualitative detection of adenovirus and its antigens  ·  Aids in the diagnosis
14 Fisher HealthCare Sure-Vue* RSV Test Kit
Alternative procedure for bloody and mucoid samples CLIA Complexity: Waived and Moderate CPT Codes: 87807QW, 87807   ·  Sensitivity >99%  ·  Specificity >99%  ·  Results in 15 minutes  ·  Specimen
15 Remel Thymune*-M Thyroid Hemagglutination Test Kit
Test and control cells treated with formalin and freeze-dried to give long-term stability on storage   ·  Haemagglutination kit for the semiquantitative measurement of auto-antibodies to human thyroid microsomal antigen  ·  CLIA Complexity: Non-waived  ·  Results
  1. 1
  2. 2
  3. 3
  4. 4
  5. 5
  6. >
  7. >>