Username:
Password:
Store:
Scientific
Safety
HealthCare
Science Education
New User?
Forgot Password?
SCIENTIFIC
SAFETY
HEALTHCARE
SCIENCE EDUCATION
Home
Help
Contact Fisher
Help Index
Site Map
Learning Center
[
close
]
Part of Thermo Fisher Scientific
Search
Submit
Substructure Search
Certificate Of Analysis
Accessories for:
BioTek* PowerWave* Microplate Spectrophotometers> PowerWave HT
Absorbance test plate
Each for $868.23
Qty:
Plate Adapter
Each for $107.71
Qty:
Cuvet; Quartz
Each for $178.14
Qty:
Shopping Cart
View Cart
No items in cart
For security reasons you will be automatically logged out after 30 minutes of inactivity.
BioTek* PowerWave* Microplate Spectrophotometers
> PowerWave HT
BT-RPRWI
Spectrophotometer, Microplate; Bio-Tek PowerWave; Scanning; HT; w/Incubation; 200-999nm; 96- and 384-well plate reading w/programmable shaking; Temp. control ambient 4 to 50°C; Includes Software
See Details
Includes:
Gen 5 data analysis software, serial cable (null modem, 9- to 95-pin female), bidirectional RS-232 interface
Item
Details
Images
Accessories
Description
Catalog Number
Quantity
Price
PowerWave HT
BT-RPRWI
Bio Tek Instruments
No.:RPRWI
Each for $20,710.00
fisherSci.com - 96/384 MICROPLATE SPEC W/INC - PowerWave HT - RPRWI