Invitrogen
Manufacturer Code:PA525964
Catalog # PIPA525964
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 65-93 amino acids from the N-terminal region of human VTN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: complement S-protein; epibolin; S-protein; serum spreading factor; Serum-spreading factor; somatomedin B; Somatomedin-B; V75; V75 VN VNT S-protein complement S-protein epibolin serum spreading factor serum-spreading factor somatomedin B; Vitronectin; Vitronectin V10 subunit; Vitronectin V65 subunit; VN
Gene Aliases: V75; VN; VNT; VTN
UniProt ID: (Human) P04004
Entrez Gene ID: (Human) 7448
Molecular Function: cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.