Invitrogen
Manufacturer Code:MA531319
Catalog # PIMA531319
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3 |
Immunogen | Recombit Human Vitronectin/VTN Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: complement S-protein; epibolin; S-protein; serum spreading factor; Serum spreading factor Complement S Protein Epibolin S Protein Somatomedin B antibody V75 VTN; Serum-spreading factor; somatomedin B; Somatomedin-B; V75; Vitronectin; Vitronectin V10 subunit; Vitronectin V65 subunit; VN
Gene Aliases: V75; VN; VNT; VTN
UniProt ID: (Human) P04004
Entrez Gene ID: (Human) 7448
Molecular Function: cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.