502 Bad Gateway
502 Bad Gateway
Sustainability Lab Equipment
Energy Efficient Freezers and Their Role in Lowering Laboratory Carbon Footprints
February 2026 | 5 min read
Energy Efficient Freezers and Their Role in Lowering Laboratory Carbon Footprints
Laboratories are among the most energy intensive environments in modern research. While high profile instruments and ventilation systems often attract attention, one of the most significant contributors to laboratory energy consumption operates continuously and often unnoticed: cold storage. Ultra low temperature freezers operating at -80⁰C can consume as much electricity as a typical household each year, a finding supported by international energy assessments of research infrastructure (International Energy Agency, 2024, “Energy Efficiency 2024”, https://www.iea.org/reports/energy-efficiency-2024).
As sustainability commitments become more prominent across academia, healthcare, and industry, laboratories are under pressure to reduce their environmental impact without compromising sample integrity or scientific rigor. Energy efficient freezers have emerged as one of the most effective ways to address this challenge.
Why Laboratory Freezers Consume So Much Energy
Ultra low temperature freezers are designed to maintain extremely stable conditions around the clock. Older models commonly rely on fixed speed compressors and comparatively inefficient insulation materials. As a result, they draw large amounts of electricity and emit substantial heat into the laboratory environment.
This excess heat increases the demand on building ventilation and air conditioning systems, creating a secondary energy burden. A detailed assessment of laboratory energy use identified cold storage as one of the largest contributors to electricity demand in biomedical research facilities, accounting for up to 10 percent of total laboratory electricity use in some institutions (My Green Lab, 2024, “The Hidden Energy Cost of Cold Storage”, https://mygreenlab.org/resources/energy).
Technological Advances in Energy Efficient Freezer Design
In response to rising energy costs and sustainability targets, freezer technology has evolved significantly. Modern ultra low temperature freezers increasingly rely on variable speed compressors that adjust cooling output based on real time demand, reducing unnecessary power consumption.
Improvements in insulation technology also play a central role. Vacuum insulated panels dramatically reduce heat transfer compared to conventional foam insulation, allowing freezers to maintain internal temperatures with less energy input. Testing under the ENERGY STAR program shows that certified ultra low temperature freezers can use up to 40 percent less energy than older, non certified models (ENERGY STAR, 2024, “ENERGY STAR Program Requirements for Laboratory Grade Freezers”, https://www.energystar.gov/products/recent_program_updates/low-temperature-freezer-technology-and-energy-efficiency).
Refrigerant choice further influences environmental impact. Traditional hydrofluorocarbon refrigerants have a high global warming potential, while newer hydrocarbon based alternatives offer improved efficiency with significantly lower climate impact, as outlined in European regulatory guidance on fluorinated greenhouse gases (European Commission, 2024, “Fluorinated Greenhouse Gases”, https://climate.ec.europa.eu/eu-action/fluorinated-greenhouse-gases_en).
Optimizing Freezer Temperature Set Points
Beyond equipment design, operational decisions strongly affect energy use. One of the most impactful measures is adjusting ultra low temperature freezer set points from -80⁰C to -70⁰C where scientifically appropriate.
Multiple studies confirm that many biological samples, including DNA, RNA, and proteins, remain stable at -70⁰C for long term storage. A peer-reviewed review in Nature Methods concluded that sample integrity is maintained for most molecular biology applications at the higher temperature, while energy demand decreases substantially (ScienceDirect, 2024, "Examining the stability of viral RNA and DNA in wastewater: Effects of storage time, temperature, and freeze-thaw cycles", https://www.sciencedirect.com/science/article/pii/S0043135424007802.)
Independent studies and technical assessments of ultra-low-temperature freezers show that increasing the temperature set point (for example from –80 °C to –70 °C) can significantly lower energy consumption and reduce compressor workload, which may contribute to longer equipment service life (Thermo Fisher Scientific, 2025, "ULT freezers: Beyond the specifications", https://www.thermofisher.com/blog/anz-science-news/ult-freezers-beyond-the-specifications).
Sustainability Benefits Beyond Electricity Savings
Lower electricity use directly reduces greenhouse gas emissions, particularly in regions where electricity generation depends on fossil fuels. However, the sustainability benefits extend further.
Reduced heat output lowers the cooling load on laboratory HVAC systems, which are among the most energy intensive components of research buildings. Improved freezer reliability also reduces the risk of sample loss, preventing the need to repeat experiments that consume additional reagents, consumables, and energy.
From a lifecycle perspective, modern freezers are designed for longer service life and improved maintainability. The International Institute for Sustainable Laboratories emphasizes equipment longevity as a key factor in reducing the overall environmental footprint of research infrastructure (International Institute for Sustainable Laboratories, 2024, “Best Practices for Sustainable Laboratories”, https://www.i2sl.org/about).
Financial and Operational Considerations
Although energy efficient freezers often involve higher upfront investment, total cost of ownership analyses consistently demonstrate long term financial benefits. Reduced electricity costs, fewer breakdowns, and lower maintenance requirements typically result in payback periods of three to five years, depending on energy prices and usage patterns (Mayo Clinic, 2025, "Replacing freezers leads to energy and cost savings", https://practicegreenhealth.org/tools-and-resources/mayo-clinic-replacing-freezers-leads-energy-and-cost-savings).
For many organizations, freezer upgrades now form part of broader sustainability driven procurement strategies aligned with institutional climate targets.
Freezer Management as Part of a Broader Sustainability Strategy
Technology alone is not sufficient to achieve meaningful reductions in laboratory carbon footprints. Effective freezer management programs combine efficient equipment with better organization, routine maintenance, and user engagement.
Regular inventory audits help identify obsolete samples and underused units that can be decommissioned. Cleaning condenser filters and ensuring adequate airflow around freezers improves performance. Behavioral changes, such as minimizing door openings and organizing samples clearly, further reduce energy waste (University of Washington, 2025, "Ultra-low temperature freezers", https://sustainability.uw.edu/green-laboratory/freezers).
Energy Efficient Freezers in the Context of Sustainable Laboratories
Cold storage optimization is most effective when integrated into a broader sustainability framework that includes efficient ventilation, responsible consumables sourcing, and digital inventory management.
Energy efficient freezers therefore act as both a technical solution and a cultural entry point into more sustainable laboratory operations.
Conclusion
Energy efficient freezers show that substantial sustainability gains in laboratories are achievable without compromising scientific quality. By combining modern freezer technology with evidence based operational practices, laboratories can significantly reduce energy consumption, lower carbon emissions, and improve working conditions.
As research activity continues to expand globally, responsible cold storage management will play an increasingly important role in enabling sustainable science.