Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
15-077-67
Fisher Scientific™ Variable-Flow Chemical Transfer Pump
Precise, seamless speed control with all-day performance
Achieves all-day pulseless fluid transfer at variable rates by way of stepless “continuous-sweep” variable speed control.
Includes: 115V wall power supply
  • Direct-drive engineering maximizes power for smooth-rolling pump action
  • Pumps fluids from 0.12 to 2.2L/min. with viscosity to 200 centipoise (0.2 Pa·s) and demonstrates suction lift of 10 ft. wet
  • Tolerates fluid temperatures from -40° to +93°C (-40° to +200°F)
  • Barbed outlet ports accommodate any type of tubing with an inside diameter of 0.2 in.
  • Chemical-resistant, wetted parts are DuPont™ Delrin™, Type 304 stainless steel, Buna N, and PTFE
  • Chemical-resistant ABS case
  • PURGE/PRIME switch allows for high-speed emptying and filling
  • Reverses to expedite tube draining
View Specifications
Images for Fisher Scientific™ Variable-Flow Chemical Transfer Pump
  • 15-077-67
15-077-67
15-077-67
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
15-077-67 Chemical Transfer; Variable Flow
Each for $431.00
Check Availability
Add
to Cart
(0) in My Cart
Description
  • Direct-drive engineering maximizes power for smooth-rolling pump action
  • Pumps fluids from 0.12 to 2.2L/min. with viscosity to 200 centipoise (0.2 Pa·s) and demonstrates suction lift of 10 ft. wet
  • Tolerates fluid temperatures from -40° to +93°C (-40° to +200°F)
  • Barbed outlet ports accommodate any type of tubing with an inside diameter of 0.2 in.
  • Chemical-resistant, wetted parts are DuPont™ Delrin™, Type 304 stainless steel, Buna N, and PTFE
  • Chemical-resistant ABS case
  • PURGE/PRIME switch allows for high-speed emptying and filling
  • Reverses to expedite tube draining
Specifications & Ordering Information:
Dimensions: 8L x 4.75W x 4.5 in.H (20.3 x 12 x 11.5cm)