UserName
New Feature
You can now browse the site in English or Español.
Antibodies
Essential Tools for Detection, Quantification, and Analysis in Research and Diagnostics
Explore the Fisher Scientific antibody collection, featuring more than 1 million primary and secondary antibodies from trusted industry suppliers. Whether you’re performing ELISA, flow cytometry, Western blotting, immunohistochemistry (IHC), immunoprecipitation (IP) or multiplex assays with fluorescently conjugated antibodies, our extensive portfolio helps you find the right antibody for every application
Best Sellers
GAPDH Rabbit anti-Human, Mouse, Plant, Porcine, Rat, Polyclonal, Proteintech™
  • Best-selling GAPDH antibody for all applications (20 µL / 150 µL)
  • Proven reliability at an affordable price
  • Test sample available for evaluation
GAPDH Rabbit anti-Human, Mouse, Plant, Porcine, Rat, Polyclonal, Proteintech
Invitrogen™ Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 488
Our #1 selling secondary fluorescent antibody. Alexa Fluor dyes are among the most trusted fluorescent dyes available today. Also available in Alexa Plus variants for even more performance  in  sensitivity, brightness, and signal-to-noise ratio.
Invitrogen™ Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 488
R&D Systems™ Proteome Profiler Human Phospho-Kinase Array Kit
The Human Phospho-Kinase Array Kit offers a fast, sensitive, and cost-effective solution for simultaneously profiling phosphorylation of 37 kinases and two related proteins, eliminating the need for multiple immunoprecipitations and Western blots to efficiently analyze cellular signaling responses.
R&D Systems™ Proteome Profiler Human Phospho-Kinase Array Kit
Discover More Life Science Products and Tools
Explore our extensive range of life sciences products, including reagents, assays, antibodies, immunoassays, and more.
Related Resources
Flow Cytometry Workflow Capabilities
Flow cytometry is an important technique central to many of the fastest-growing areas of life science research. Learn more about flow cytometry.
Flow Cytometry Workflow Capabilities
Nano-Traps for Superior Immunoprecipitation
Ready-to-use beads for fast and efficient immunoprecipitation.
Nano-Traps for Superior Immunoprecipitation
Better Images with Nanobodies
Nano-secondaries, nano-boosters, nano-labels, chromobodies, and nanobody-fc fusions for immunofluorescence.
Better Images with Nanobodies
Other Applications
Cell Analysis Methods
Find what you need for flow cytometry, cell imaging, and more.
Cell Analysis Methods
Life Sciences Products
Explore products for cell, molecular and protein biology, and more.
Life Sciences Products
Fisherbrand Products
Value-packed products that make your lab life easier - guaranteed.
Fisherbrand Products
Antibodies FAQ for Life Science Research
Antibodies are specialized proteins—also known as immunoglobulins—to recognize and bind to specific antigens, such as bacteria, viruses, or other foreign molecules. Antibodies are used in life science research to detect, isolate, and quantify specific molecules. In research and diagnostics, antibodies are used as highly specific tools to detect, quantify, or isolate target proteins or molecules. Depending on their design, they can be primary antibodies (binding directly to the target) or secondary antibodies (binding to the primary antibody to enhance detection).
Yes, antibodies are glycoproteins with a defined antibody structure consisting of variable and constant regions. This structure enables precise binding to target antigens, making antibodies indispensable in molecular biology, cancer research, and diagnostics.
In research, antibodies are used to:
  • Detect proteins and other biomolecules
  • Visualize cellular components
  • Purify specific targets
  • Track biological processes
This includes both direct fluorescent antibody and indirect fluorescent antibody techniques, which rely on antibody–antigen interactions for signal detection. These methods are widely used in cancer biology and immunological studies.
Antibodies bind to specific epitopes on antigens. In assays like ELISA, Western blot, and Flow Cytometry, this binding is linked to a detectable signal—such as fluorescence or enzymatic activity (e.g., HRP-conjugated antibodies)—allowing researchers to measure the presence and quantity of the target molecule.
Monoclonal antibodies are identical antibodies derived from a single B-cell clone. When researchers ask “what are monoclonal antibodies?”, they’re referring to highly specific reagents used in diagnostics, cancer therapy, and analytical assays.
Monoclonal antibodies bind to a single epitope with high specificity. This makes them ideal for applications requiring reproducibility, such as biomarker detection, target validation, and cell sorting in cancer and immunology research.
Antibodies are produced by immunizing animals (e.g., mice or rabbits) with a target antigen. The resulting antibody–antibody interactions are studied and refined for use in research. Recombinant technologies also allow for custom antibody production, including Alexa-conjugated and HRP-conjugated formats.
Monoclonal antibodies are created by fusing B-cells with myeloma cells to form hybridomas. These are screened for specificity and cultured to produce large quantities of uniform antibodies used in cancer diagnostics and therapeutic development.
Secondary antibodies bind to primary antibodies and are often conjugated with enzymes like HRP or fluorophores such as Alexa Fluor dyes. They are used in indirect fluorescent antibody techniques to amplify signals in assays like ELISA, Western blot, Flow Cytometry, and Immunohistochemistry (ICH), enhancing sensitivity and specificity in cancer and cell biology research.
An ANA antibody (antinuclear antibody) targets nuclear components and is used in research to study autoimmune diseases. It’s commonly detected using indirect fluorescent antibody assays and is relevant in both immunology and cancer-related autoimmunity studies.
High TPO antibodies (thyroid peroxidase antibodies) are markers of autoimmune thyroid conditions like Hashimoto’s thyroiditis. In research, they are studied as part of thyroid antibody profiles but are not direct indicators of cancer. However, they may be included in broader immunological panels in cancer-related studies.
Featured Key Antibody Suppliers
The Fisher Scientific channel brings you antibodies from top global suppliers delivering high-quality, reliable antibodies for accurate and reproducible results across all research applications.