Gene Transcripts

Gene Symbol
CYP4F3
Entrez Gene ID
4051
Gene Name
cytochrome P450 family 4 subfamily F member 3
Gene Aliases
CPF3, CYP4F, LTB4H
Chromosome Location
Chr. 19: 15640897 - 15660760 on Build GRCh38
UniGene ID
Hs.106242
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000896.2 NP_000887.2 3 263
XM_011528014.2 XP_011526316.1 1 15
GenBank mRNA AB002454.1 - 3 263
AK298675.1 - 3 243
D12620.1 - 3 247
D12621.1 - 3 229

View Full Product Details