Gene Transcripts
- Gene Symbol
- KIAA0087
- Entrez Gene ID
- 9808
- Gene Name
- KIAA0087
- Gene Aliases
- Chromosome Location
- Chr. 7: 26533121 - 26538825 on Build GRCh38
- UniGene ID
- Hs.69749
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_022006.1 |
|
2 |
2441 |
|
GenBank mRNA |
D42038.1 |
- |
2 |
2403 |
|
View Full Product Details