This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00017060_cn
See other KCNE3 CNV Assays ›
Gene Symbol
KCNE3
Assay Reference Genome
Location

Chr.11:74456918 on build GRCh38
Cytoband
11q13.4
Assay ID Hs00017060_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

10008

Gene Name:

potassium voltage-gated channel subfamily E regulatory subunit 3

Gene Aliases:

HOKPP, HYPP, MiRP2

Location:

Chr.11:74454841-74467729 on Build GRCh38

Assay Gene Location:

Within Exon 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
KCNE3 NM_005472.4 3 993 NP_005463.1
XM_011544713.2 2 843 XP_011543015.1
XM_017017047.1 4 1063 XP_016872536.1
XM_017017048.1 5 1159 XP_016872537.1
XM_017017049.1 3 1009 XP_016872538.1
XM_017017050.1 3 3867 XP_016872539.1
XM_017017051.1 4 982 XP_016872540.1
XM_017017052.1 3 933 XP_016872541.1
AF302494.1 2 951 AAG16255.1
BC110612.2 2 762 AAI10613.1
BC113743.1 2 817 AAI13744.1
CR936625.1 3 987 CAI56768.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1041154 Chr.11:74403756 - 74466864 on Build GRCh38 Loss KCNE3
esv34979 Chr.11:74167643 - 74822407 on Build GRCh38 Gain CHRDL2 MIR548AL KCNE3 C2CD3 PGM2L1 RNF169 POLD3 LOC100287896 P4HA3 PPME1 MIR4696 LIPT2 LOC101928580
esv3626840 Chr.11:74371082 - 74464389 on Build GRCh38 Gain MIR548AL KCNE3 PGM2L1
esv2759837 Chr.11:74131564 - 74846552 on Build GRCh38 Gain CHRDL2 MIR548AL KCNE3 C2CD3 PGM2L1 RNF169 POLD3 LOC100287896 XRRA1 P4HA3 PPME1 MIR4696 LIPT2 LOC101928580
nsv555443 Chr.11:74398121 - 74469444 on Build GRCh38 Loss MIR548AL KCNE3 PGM2L1
nsv832209 Chr.11:74444838 - 74618074 on Build GRCh38 Gain LOC100287896 KCNE3 LIPT2 POLD3
dgv209e214 Chr.11:74225600 - 74828943 on Build GRCh38 Gain LOC100287896 CHRDL2 P4HA3 MIR548AL KCNE3 PPME1 MIR4696 LIPT2 PGM2L1 RNF169 POLD3 LOC101928580
nsv393 Chr.11:74424817 - 74470372 on Build GRCh38 Deletion KCNE3
nsv825994 Chr.11:74225622 - 74823410 on Build GRCh38 Gain LOC100287896 CHRDL2 P4HA3 MIR548AL KCNE3 PPME1 MIR4696 LIPT2 PGM2L1 RNF169 POLD3 LOC101928580

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

voltage-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

regulation of membrane repolarization
potassium ion transmembrane transport
positive regulation of voltage-gated calcium channel activity
negative regulation of delayed rectifier potassium channel activity
voltage-gated potassium channel activity
protein binding
potassium channel regulator activity
ion channel binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0